Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betahistine Hydrochloride, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18600933 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-933 1 kg Amber Glass Bottle
18-600-930 5 g Amber Glass Bottle
18-600-931 25 g Amber Glass Bottle
18-600-932 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-600-933 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B19761KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

B1976, 5579-84-0, C8H12N2·2HCl

Spectrum™ Chemical Betahistine Hydrochloride, USP is an anti-vertigo drug. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Identifiants chimiques

CAS 5579-84-0
Molecular Formula C8H14Cl2N2
Molecular Weight (g/mol) 209.11
InChI Key XVDFMHARQUBJRE-UHFFFAOYSA-N
IUPAC Name dihydrogen methyl[2-(pyridin-2-yl)ethyl]amine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].CNCCC1=CC=CC=N1

Spécifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 99 to 101%
Grade USP
Physical Form Crystalline Powder or Lumps
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.