accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55804-65-4

IUPAC Name: 4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carboxylate
Synonyms: 4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carboxylate
Molecular Weight (g/mol): 284.29
Molecular Formula: C16H14NO4
InChi Key: KCDCNGXPPGQERR-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC2=CC3=C4N(CCC3)CCCC4=C2OC1=O
Quantity 
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results