Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin 343, TCI America™
SDP

Catalog No. C2899200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2899200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2899200MG Supplier TCI America Supplier No. C2899200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 55804-65-4
Molecular Formula C16H15NO4
Molecular Weight (g/mol) 285.299
MDL Number MFCD00051335
InChI Key KCDCNGXPPGQERR-UHFFFAOYSA-N
Synonym coumarin 343, 11-oxo-2,3,5,6,7,11-hexahydro-1h-pyrano 2,3-f pyrido 3,2,1-ij quinoline-10-carboxylic acid, 1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizine-10-carboxylicacid, 2,3,6,7-tetrahydro-11-oxo, 2,3,6,7-tetrahydro-11-oxo-1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizine-10-carboxylic acid, 1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizine-10-carboxylic acid, 2,3,6,7-tetrahydro-11-oxo, coumarin343, 11-oxo-2,3,6,7-tetrahydro-1h,5h,11h-pyrano 2,3-f pyrido 3,2,1-ij quinoline-10-carboxylic acid, coumarin-343
PubChem CID 108770
ChEBI CHEBI:51941
SMILES C1CC2=C3C(=C4C(=C2)C=C(C(=O)O4)C(=O)O)CCCN3C1

Specifications

Melting Point 235°C
Quantity 200 mg
Formula Weight 285.30
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 343
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.