accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONOO

CAS RN 55804-66-5

IUPAC Name: ethyl 4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carboxylate
Synonyms: coumarin 314 coumarin 504
Molecular Weight (g/mol): 313.35
Molecular Formula: C18H19NO4
InChi Key: VMJKUPWQKZFFCX-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC3=C4N(CCC3)CCCC4=C2OC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)