Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin 314, Laser grade, MP Biomedicals™

Catalog No. ICN19036183 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19036183 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19036183 Supplier MP Biomedicals, Inc Supplier No. 0219036183

Chemical Identifiers

CAS 55804-66-5
Molecular Formula C18H19NO4
Molecular Weight (g/mol) 313.353
InChI Key VMJKUPWQKZFFCX-UHFFFAOYSA-N
PubChem CID 72653
ChEBI CHEBI:51940
SMILES CCOC(=O)C1=CC2=CC3=C4C(=C2OC1=O)CCCN4CCC3

Specifications

Formula Weight 313.4
Grade Laser
Color Yellow
Quantity 250 mg
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 314
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.