accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55804-67-6

IUPAC Name: 5-acetyl-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Synonyms: 5-acetyl-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Molecular Weight (g/mol): 283.33
Molecular Formula: C17H17NO3
InChi Key: JBPCDMSEJVCNGV-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC2=CC3=C4N(CCC3)CCCC4=C2OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results