Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin 334, 96%

Catalog No. AAH33332ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33332ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH33332ME Supplier Thermo Scientific Chemicals Supplier No. H33332ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 55804-67-6 | C17H17NO3 | 283.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55804-67-6
Molecular Formula C17H17NO3
Molecular Weight (g/mol) 283.33
MDL Number MFCD00051336
InChI Key JBPCDMSEJVCNGV-UHFFFAOYSA-N
Synonym coumarin 334, 9-acetyl-2,3,5,6-tetrahydro-1h,4h-11-oxa-3a-aza-benzo de anthracen-10-one, 1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizin-11-one, 10-acetyl-2,3,6,7-tetrahydro, 10-acetyl-2,3,6,7-tetrahydro-1h,5h,11h-pyrano-2,3-f pyrido 3,2,1-ij quinolin-11-one, 10-acetyl-2,3,6,7-tetrahydro-1h,5h,11h-pyrano 2,3-f pyrido 3,2,1-ij quinolin-11-one, coumarin-334, coumarin 521, coumarin 334, dye content 99 %, 3-acetyl-6,7,9,10-tetrahydro-8h,11h-quinolizino 9,9a, 1-gh coumarin
PubChem CID 72655
IUPAC Name 5-acetyl-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
SMILES CC(=O)C1=CC2=CC3=C4N(CCC3)CCCC4=C2OC1=O

Specifications

Melting Point 176°C to 181°C
Quantity 500 mg
UN Number UN2811
Formula Weight 448.53
Percent Purity 96%
Chemical Name or Material Coumarin 334
Hazard Category H302+H312+H332-H315-H319-H335-H500
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 259-826-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.