accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55894-52-5

IUPAC Name: 4-nitro-3-(octanoyloxy)benzoic acid
Synonyms: 4-nitro-3-(octanoyloxy)benzoic acid
Molecular Weight (g/mol): 309.32
Molecular Formula: C15H19NO6
InChi Key: VDCATWRTRISVBZ-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)OC1=C(C=CC(=C1)C(O)=O)[N+]([O-])=O
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results