accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFF

CAS RN 559-40-0

IUPAC Name: octafluorocyclopent-1-ene
Synonyms: cyclopentene, octafluoro-
Molecular Weight (g/mol): 212.04
Molecular Formula: C5F8
InChi Key: YBMDPYAEZDJWNY-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)(F)C(F)(F)C1(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results