accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO(R)OOCH3(S)H3C(S)(S)O(S)H3C(R)F(S)OH(S)H3C

CAS RN 5593-20-4

IUPAC Name: 2-[(1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1-(propanoyloxy)-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl propanoate
Synonyms: betamethasone dipropionate
Molecular Weight (g/mol): 504.6
Molecular Formula: C28H37FO7
InChi Key: CIWBQSYVNNPZIQ-XYWKZLDCSA-N
SMILES: CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
Grade 
  • (1)
  • (5)
Industry Type 
  • (5)
Percent Purity 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (5)
  • (5)
  • (1)
  • (5)
  • (5)
  • (5)