Learn More
Betamethasone Dipropionate, Micronized Powder, USP, 97-103%, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
Spectrum™ Chemical Betamethasone Dipropionate, Micronized Powder, USP is a glucocorticoid steroid applied as a topical cream, ointment, lotion or gel to treat itching and other minor skin conditions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| CAS Max % | 100% |
| Industry Type | Pharmaceutical |
| Quantity | 100 g |
| Percent Purity | 97 to 103% |
| Grade | USP |
| Chemical Name or Material | Betamethasone Dipropionate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.