CAS RN 55985-85-8
IUPAC Name:
(2-carboxyethyl)triphenylphosphanium methane bromide
Synonyms:
(2-carboxyethyl)triphenylphosphanium methane bromide
Molecular Weight (g/mol):
431.31
Molecular Formula:
C22H24BrO2P
InChi Key:
MBIJNKFEWFACKA-UHFFFAOYSA-N
SMILES:
C.[Br-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
(2-Carboxyethyl)triphenylphosphonium tribromide, 98+%, Thermo Scientific™
CAS: 55985-85-8 Molecular Formula: C22H24BrO2P Molecular Weight (g/mol): 431.31 MDL Number: MFCD00051883 InChI Key: MBIJNKFEWFACKA-UHFFFAOYSA-N Synonym: 2-carboxyethyl triphenylphosphanium tribromide PubChem CID: 57350967 IUPAC Name: 2-carboxyethyl(triphenyl)phosphanium;tribromide SMILES: C.[Br-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1