Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Carboxyethyl)triphenylphosphonium tribromide, 98+%, Thermo Scientific™

Catalog No. AAL0055306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0055306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0055306 Supplier Thermo Scientific Chemicals Supplier No. L0055306

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(2-Carboxyethyl)triphenylphosphonium tribromide, is a fine chemical and pharmaceutical intermediate. It is a bromine transfer agent for selective α-bromination of ketones and ketone acetals in the presence of double bonds, activated aromatic rings, esters or another free enolic position.

Solubility
It reacts with water. Soluble in THF (25 g/100 mL), dichloromethane, insoluble in alcohol.

Notes
Moisture, water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. This reagent should be handled in a fume hood.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55985-85-8
Molecular Formula C22H24BrO2P
Molecular Weight (g/mol) 431.31
MDL Number MFCD00051883
InChI Key MBIJNKFEWFACKA-UHFFFAOYSA-N
Synonym 2-carboxyethyl triphenylphosphanium tribromide
PubChem CID 57350967
IUPAC Name 2-carboxyethyl(triphenyl)phosphanium;tribromide
SMILES C.[Br-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∽159°C (decomposition)
Quantity 5 g
UN Number UN3261
Beilstein 6262851
Sensitivity Moisture sensitive
Solubility Information It reacts with water. Soluble in THF (25g/100 mL),dichloromethane,insoluble in alcohol.
Formula Weight 575.09
Percent Purity ≥98%
Chemical Name or Material (2-Carboxyethyl)triphenylphosphonium tribromide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.