accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NOHNOH

CAS RN 56-09-7

IUPAC Name: 2-amino-6-hydroxy-3,4-dihydropyrimidin-4-one
Synonyms: 2-amino-6-hydroxy-3H-pyrimidin-4-one
Molecular Weight (g/mol): 127.1
Molecular Formula: C4H5N3O2
InChi Key: AUFJTVGCSJNQIF-UHFFFAOYSA-N
SMILES: NC1=NC(O)=CC(=O)N1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (3)
  • (4)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (4)
  • (2)
  • (5)
  • (1)
  • (6)
  • (3)
  • (4)
  • (1)
  • (2)