accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56-17-7

IUPAC Name: dihydrogen 2-[(2-aminoethyl)disulfanyl]ethan-1-amine dichloride
Synonyms: cystamine
Molecular Weight (g/mol): 225.19
Molecular Formula: C4H14Cl2N2S2
InChi Key: YUFRRMZSSPQMOS-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCCSSCCN
Molecular Weight (g/mol) 
  • (7)
  • (8)
Quantity 
  • (2)
  • (5)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (3)
  • (5)
  • (7)
  • (8)
  • (2)
  • (5)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)