Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dithiobis(ethylamine)Di HCl, MP Biomedicals

Catalog No. ICN212984 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN212984 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN212984 Supplier MP Biomedicals, Inc Supplier No. 0521298401

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56-17-7
Molecular Formula C4H14Cl2N2S2
Molecular Weight (g/mol) 225.19
InChI Key YUFRRMZSSPQMOS-UHFFFAOYSA-N
Synonym cystamine dihydrochloride, cystamin dihydrochloride, ethanamine, 2,2'-dithiobis-, dihydrochloride, usaf cb-34, 2,2'-disulfanediyldiethanamine dihydrochloride, 2-aminoethyl disulfide dihydrochloride, bis 2-aminoethyl disulfide dihydrochloride, cystamine 2hcl, 2,2'-dithiodi ethylammonium dichloride, decarboxycystine dihydrochloride
PubChem CID 5941
IUPAC Name 2-(2-aminoethyldisulfanyl)ethanamine;dihydrochloride
SMILES C(CSSCCN)N.Cl.Cl

Specifications

Quantity 1 g
Formula Weight 225.21
Chemical Name or Material 2, 2-Dithiobis(Ethylamine)Di HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.