accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NHNNH2

CAS RN 56-18-8

IUPAC Name: bis(3-aminopropyl)amine
Synonyms: norspermidine 3,3'-iminodi(propylamine) 3,3-diaminodipropylamine propylamine, 3,3'-iminobis- dipropylamine, 3,3'-diamino- 3,3'-iminobispropylamine 3,3'-iminobis(propylamine) 3,3'-iminobis-propylamine 3,3'-azanediyldi(propanamine) imino-bis(3-propylamine)
Molecular Weight (g/mol): 131.22
Molecular Formula: C6H17N3
InChi Key: OTBHHUPVCYLGQO-UHFFFAOYSA-N
SMILES: NCCCNCCCN
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)