Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(3-aminopropyl)amine

Catalog No. I1006500GA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I1006500GA 500 g
I1006100GA 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I1006500GA Supplier Sigma Aldrich Supplier No. I1006500GA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(3-aminopropyl)amine

Specifications

CAS 56-18-8
Melting Point -14°C (lit.)
Density 0.938 g/mL (at 25°C (literature))
Boiling Point 151°C (50 mmHg)
Molecular Formula C6H17N3
Refractive Index n20/D 1.481 (literature)
Linear Formula (NH2CH2CH2CH2)2 NH
MDL Number MFCD00008214
Quantity 500 g
Synonym BAPA; 3,3 -Diaminodipropylamine; 3,3 -Iminodipropylamine; N-(3-Aminopropyl)-1,3-propanediamine; Dipropylenetriamine; Norspermidine
Molecular Weight (g/mol) 131.22
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.