CAS RN 56-35-9
IUPAC Name:
tributyltin oxide
Synonyms:
tributyltin oxide
Molecular Weight (g/mol):
600.15
Molecular Formula:
C24H58OSn2
InChi Key:
NZQFYJHTWVEUPP-UHFFFAOYSA-N
SMILES:
O.CCCC[SnH](CCCC)CCCC.CCCC[SnH](CCCC)CCCC
1
–
1
of
1
results
1
Promotions Available
Bis(tri-n-butyltin) oxide, 97%
Bis(tri-n-butyltin) oxide, 97%
CAS: 56-35-9 Molecular Formula: C24H58OSn2 Molecular Weight (g/mol): 600.15 MDL Number: MFCD00009418 InChI Key: NZQFYJHTWVEUPP-UHFFFAOYSA-N Synonym: tributyltin oxide,tbto,bis tributyltin oxide,hexabutyldistannoxane,distannoxane, hexabutyl,lastanox q,bis tri-n-butyltin oxide,butinox,biomet PubChem CID: 16682746 ChEBI: CHEBI:81543 IUPAC Name: tributyl(tributylstannyloxy)stannane SMILES: O.CCCC[SnH](CCCC)CCCC.CCCC[SnH](CCCC)CCCC