Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(tri-n-butyltin) oxide, 97%

Catalog No. AAA1324222
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1324222 100 g
AAA1324236 500 g
AAA132420E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1324222 Supplier Thermo Scientific Chemicals Supplier No. A1324222
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 56-35-9 | C24H58OSn2 | 600.15 g/mol

Bis(tri-n-butyltin) oxide is employed in the synthesis of α,β-unsaturated methyl ketones, isoxazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(tri-n-butyltin) oxide is employed in the synthesis of α,β-unsaturated methyl ketones, isoxazoles.

Solubility
Soluble in water (0.071 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-35-9
Molecular Formula C24H58OSn2
Molecular Weight (g/mol) 600.15
MDL Number MFCD00009418
InChI Key NZQFYJHTWVEUPP-UHFFFAOYSA-N
Synonym tributyltin oxide, tbto, bis tributyltin oxide, hexabutyldistannoxane, distannoxane, hexabutyl, lastanox q, bis tri-n-butyltin oxide, butinox, biomet
PubChem CID 16682746
ChEBI CHEBI:81543
IUPAC Name tributyl(tributylstannyloxy)stannane
SMILES O.CCCC[SnH](CCCC)CCCC.CCCC[SnH](CCCC)CCCC

Specifications

Melting Point -45°C
Density 1.172
Boiling Point 179°C to 180°C (2 mmHg)
Flash Point 168°C (334°F)
Odor Irritating
Refractive Index 1.4865
Linear Formula [(CH3CH2CH2CH2)3Sn]2O
Quantity 100 g
UN Number UN2788
Beilstein 745057
Solubility Information Soluble in water (0.071g/L)
Formula Weight 596.07
Percent Purity 97%
Chemical Name or Material Bis(tri-n-butyltin) oxide
Show More Show Less
Hazard Category H301-H312-H315-H319-H335-H360FD-H372
Hazard Statement GHS H Statement
H301-H311-H360-H372-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 200-268-0
RTECSNumber JN8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.