accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSnHOHH3COH3CH3C

CAS RN 56-36-0

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 351.12
Molecular Formula: C14H32O2Sn
InChi Key: MQLOAMURDGMDHF-UHFFFAOYSA-N
SMILES: CC(O)=O.CCCC[SnH](CCCC)CCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results