Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyltin Acetate, TCI America™
SDP

Catalog No. T067725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T067725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T067725G Supplier TCI America Supplier No. T067725G

Chemical Identifiers

CAS 56-36-0
Molecular Formula C14H30O2Sn
Molecular Weight (g/mol) 349.102
MDL Number MFCD00015228
InChI Key PWBHRVGYSMBMIO-UHFFFAOYSA-M
Synonym tri-n-butyltin acetate, tributyltin acetate, acetoxytributyltin, tribuyltin acetate, acetoxytributylstannane, tributylacetoxystannane, tin, acetoxytributyl, stannane, acetyloxy tributyl, stannane, acetoxytributyl, tributylstannium acetate
PubChem CID 16682741
IUPAC Name tributylstannyl acetate
SMILES CCCC[Sn](CCCC)(CCCC)OC(=O)C

Specifications

Boiling Point 120°C
Melting Point 85°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
UN Number 3146
Formula Weight 349.10
Percent Purity ≥95.0% (W)
Chemical Name or Material Tributyltin Acetate
EINECSNumber (2)-2245&(2)-2264
RTECSNumber WH5775000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.