accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPSOCH3OClOOCH3

CAS RN 56-72-4

IUPAC Name: O-3-chloro-4-methyl-2-oxo-2H-chromen-7-yl O,O-diethyl phosphorothioate
Synonyms: meldane caswell no. 335 C14H16ClO5PS coumafos CO-ral coumaphos (substance) suntol meldone resitox baymix 50
Molecular Weight (g/mol): 362.76
Molecular Formula: C14H16ClO5PS
InChi Key: BXNANOICGRISHX-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)OC1=CC=C2C(C)=C(Cl)C(=O)OC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results