CAS RN 56001-37-7
Guanosine-5'-Triphosphate Disodium Salt, MP Biomedicals™
CAS: 56001-37-7 Molecular Formula: C10H14N5Na2O14P3 Molecular Weight (g/mol): 567.14 MDL Number: MFCD03410297 InChI Key: FIZIYLKEXVIRHJ-KHRSEZDTNA-L Synonym: guanosine 5'-triphosphate,guanosine-5'-triphosphate disodium salt dihydrate gtp,guanosine-triphosphate,guanosine 5'-triphosphate 4-,gtp 4-,2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate,2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate PubChem CID: 131676145 IUPAC Name: disodium;(2R,3R,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate SMILES: [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1
Thermo Scientific Chemicals Guanosine-5'-triphosphate disodium salt
CAS: 56001-37-7 Molecular Formula: C10H14N5Na2O14P3 Molecular Weight (g/mol): 567.14 MDL Number: MFCD03410297 InChI Key: FIZIYLKEXVIRHJ-KHRSEZDTNA-L Synonym: guanosine 5'-triphosphate,guanosine-5'-triphosphate disodium salt dihydrate gtp,guanosine-triphosphate,guanosine 5'-triphosphate 4-,gtp 4-,2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate,2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate PubChem CID: 131676145 IUPAC Name: disodium;(2R,3R,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate SMILES: [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1
Bismuth Lead Tin ingot (Rose's metal)
CAS: 56001-37-7 Molecular Formula: C10H14N5Na2O14P3 Molecular Weight (g/mol): 567.14 MDL Number: MFCD03410297 InChI Key: FIZIYLKEXVIRHJ-KHRSEZDTNA-L Synonym: guanosine 5'-triphosphate,guanosine-5'-triphosphate disodium salt dihydrate gtp,guanosine-triphosphate,guanosine 5'-triphosphate 4-,gtp 4-,2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate,2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate PubChem CID: 131676145 IUPAC Name: ({[(2R,3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(phosphonatooxy)phosphinate SMILES: [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1
Thermo Scientific Chemicals Guanosine 5'-triphosphate, disodium salt hydrate, 90%, for biochemistry
CAS: 56001-37-7 Molecular Formula: C10H14N5Na2O14P3 Molecular Weight (g/mol): 567.14 MDL Number: MFCD03410297 InChI Key: FIZIYLKEXVIRHJ-KHRSEZDTNA-L Synonym: guanosine 5'-triphosphate,guanosine-5'-triphosphate disodium salt dihydrate gtp,guanosine-triphosphate,guanosine 5'-triphosphate 4-,gtp 4-,2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate,2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate PubChem CID: 131676145 IUPAC Name: disodium;(2R,3R,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate SMILES: [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1
Guanosine-5'-triphosphate, disodium, 98.1%, MP Biomedicals™
CAS: 56001-37-7 Molecular Formula: C10H14N5Na2O14P3 Molecular Weight (g/mol): 567.14 MDL Number: MFCD03410297 InChI Key: FIZIYLKEXVIRHJ-KHRSEZDTNA-L Synonym: guanosine 5'-triphosphate,guanosine-5'-triphosphate disodium salt dihydrate gtp,guanosine-triphosphate,guanosine 5'-triphosphate 4-,gtp 4-,2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate,2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate PubChem CID: 131676145 IUPAC Name: disodium phosphono ({[(2R,3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate SMILES: [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1