accessibility menu, dialog, popup

UserName

Viewing profile as user:
2H2H2H2H2H2H2H2H2H2H

CAS RN 56004-61-6

IUPAC Name: di(²H₃)methyl(²H₄)benzene
Synonyms: di(²H₃)methyl(²H₄)benzene
Molecular Weight (g/mol): 116.23
Molecular Formula: C8H10
InChi Key: CTQNGGLPUBDAKN-ZGYYUIRESA-N
SMILES: [2H]C1=C([2H])C([2H])=C(C(=C1[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results