Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Xylene-d{10}, 98+% (Isotopic)

Catalog No. AA3647206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3647206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3647206 Supplier Thermo Scientific Chemicals Supplier No. 03647206
  • $323.00
    $265.65 / Each of 1

    Save $57.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 56004-61-6 | C8H10 | 116.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56004-61-6
Molecular Formula C8H10
Molecular Weight (g/mol) 116.23
MDL Number MFCD00037698
InChI Key CTQNGGLPUBDAKN-ZGYYUIRESA-N
Synonym o-xylene-d10, 2h10-o-xylene, 1,2-dimethylbenzene-d10 99.5 atom % d, 1,2-xylene-d10, o-xylene-d10, 99 atom % d, bis 2 h? methyl 2 h? benzene, 1,2,3,4-tetradeuterio-5,6-bis trideuteriomethyl benzene
PubChem CID 91976
IUPAC Name 1,2,3,4-tetradeuterio-5,6-bis(trideuteriomethyl)benzene
SMILES [2H]C1=C([2H])C([2H])=C(C(=C1[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Sensitivity Moisture
Color Colorless
Quantity 5 g
Density 0.953 g/mL
Melting Point -25°C
Boiling Point 143°C to 145°C
UN Number UN1307
Merck Index 14,10081
Formula Weight 116.25
Percent Purity ≥98%
Chemical Name or Material o-Xylene-d{10}, Isotopic
Show More Show Less
Hazard Category H226-H312+H332-H315-H319
Hazard Statement GHS H Statement
H226-H312-H332-H315
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 259-942-8
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.