accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 560079-02-9

IUPAC Name: 2-methoxy-N,N-bis(2-methylpropyl)benzamide
Synonyms: 2-methoxy-N,N-bis(2-methylpropyl)benzamide
Molecular Weight (g/mol): 263.38
Molecular Formula: C16H25NO2
InChi Key: BQMSFQBLAFUJTK-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)N(CC(C)C)CC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results