accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3HO

CAS RN 56024-44-3

IUPAC Name: methyl 3-(4-hydroxy-3-methoxyphenyl)propanoate
Synonyms: methyl 3-(4-hydroxy-3-methoxyphenyl)propanoate
Molecular Weight (g/mol): 210.23
Molecular Formula: C11H14O4
InChi Key: PDTCYIZPTRRYOT-UHFFFAOYSA-N
SMILES: COC(=O)CCC1=CC(OC)=C(O)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results