Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-(4-Hydroxy-3-methoxyphenyl)propionate 98.0+%, TCI America™
SDP

Catalog No. M18035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18035G 5 g
M180325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18035G Supplier TCI America Supplier No. M18035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56024-44-3
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.229
InChI Key PDTCYIZPTRRYOT-UHFFFAOYSA-N
Synonym 3-(4-Hydroxy-3-methoxyphenyl)propionic Acid Methyl Ester
PubChem CID 523498
IUPAC Name methyl 3-(4-hydroxy-3-methoxyphenyl)propanoate
SMILES COC1=C(C=CC(=C1)CCC(=O)OC)O

Specifications

Color Yellow
Boiling Point 146°C
Quantity 5 g
Formula Weight 210.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-(4-Hydroxy-3-methoxyphenyl)propionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.