accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 5617-41-4

IUPAC Name: heptylcyclohexane
Synonyms: heptylcyclohexane
Molecular Weight (g/mol): 182.35
Molecular Formula: C13H26
InChi Key: MSTLSCNJAHAQNU-UHFFFAOYSA-N
SMILES: CCCCCCCC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results