Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptylcyclohexane 99.0+%, TCI America™
SDP

Catalog No. H044225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H044225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H044225ML Supplier TCI America Supplier No. H044225ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5617-41-4
Molecular Formula C13H26
Molecular Weight (g/mol) 182.351
MDL Number MFCD00019404
InChI Key MSTLSCNJAHAQNU-UHFFFAOYSA-N
Synonym 1-Cyclohexylheptane
PubChem CID 21832
IUPAC Name heptylcyclohexane
SMILES CCCCCCCC1CCCCC1

Specifications

Color Colorless
Boiling Point 244°C
Quantity 25 mL
Formula Weight 182.35
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.