accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3COH3CNHOOOHO

CAS RN 562-10-7

IUPAC Name: butanedioic acid; dimethyl({2-[1-phenyl-1-(pyridin-2-yl)ethoxy]ethyl})amine
Synonyms: pyridoxine/doxylamine
Molecular Weight (g/mol): 388.46
Molecular Formula: C21H28N2O5
InChi Key: KBAUFVUYFNWQFM-UHFFFAOYNA-N
SMILES: OC(=O)CCC(O)=O.CN(C)CCOC(C)(C1=CC=CC=C1)C1=CC=CC=N1
Grade 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (4)
Physical Form 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (4)
  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)