Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxylamine succinate, 99.46%, MP Biomedicals™

Catalog No. ICN15101705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15101705 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15101705 Supplier MP Biomedicals, Inc Supplier No. 0215101705

Chemical Identifiers

CAS 562-10-7
Molecular Formula C21H28N2O5
Molecular Weight (g/mol) 388.46
MDL Number MFCD00056168
InChI Key KBAUFVUYFNWQFM-UHFFFAOYNA-N
Synonym doxylamine succinate, decapryn, evigoa d, a-h injection, decapryn succinate, unisom, alsadorm, gittalun, mereprine
PubChem CID 11224
ChEBI CHEBI:82461
IUPAC Name butanedioic acid; dimethyl({2-[1-phenyl-1-(pyridin-2-yl)ethoxy]ethyl})amine
SMILES OC(=O)CCC(O)=O.CN(C)CCOC(C)(C1=CC=CC=C1)C1=CC=CC=N1

Specifications

Color White to Yellow
Quantity 5 g
Formula Weight 388.5
Percent Purity 0.9946
Physical Form Crystals
Chemical Name or Material Doxylamine succinate
EINECSNumber 209-228-7
Recommended Storage Store at 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.