accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COO

CAS RN 562-46-9

IUPAC Name: 4,4-dimethylcyclohexane-1,3-dione
Synonyms: 4,4-dimethylcyclohexane-1,3-dione
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: PLGPBTCNKJQJHQ-UHFFFAOYSA-N
SMILES: CC1(C)CCC(=O)CC1=O
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)