Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Dimethyl-1,3-cyclohexanedione 97.0+%, TCI America™
SDP

Catalog No. D26815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26815G Supplier TCI America Supplier No. D26815G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 562-46-9
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00075224
InChI Key PLGPBTCNKJQJHQ-UHFFFAOYSA-N
Synonym 4,4-dimethyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 4,4-dimethyl, 4,4-dimethyl-cyclohexane-1,3-dione, acmc-209lre, 4,4-dimethyl-1,3cyclohexanedione, 4,4dimethyl-1,3-cyclohexanedione, 4,4-dimethy 1,3-cyclohexanedione, 1,3-cyclohexanedione,4,4-dimethyl, 4,4-dimethyl-1,3-cyclohexanedione #
PubChem CID 136360
IUPAC Name 4,4-dimethylcyclohexane-1,3-dione
SMILES CC1(C)CCC(=O)CC1=O

Specifications

Melting Point 103°C
Color White-Yellow
Boiling Point 130°C
Quantity 5 g
Formula Weight 140.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4-Dimethyl-1,3-cyclohexanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.