accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CH3C

CAS RN 562-49-2

IUPAC Name: 3,3-dimethylpentane
Synonyms: 3,3-dimethylpentane
Molecular Weight (g/mol): 100.21
Molecular Formula: C7H16
InChi Key: AEXMKKGTQYQZCS-UHFFFAOYSA-N
SMILES: CCC(C)(C)CC
Molecular Weight (g/mol) 
  • (5)
  • (1)
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)