accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3COHNOHOH2N

CAS RN 56217-82-4

IUPAC Name: 2-(2-amino-4-methylpentanamido)-3-phenylpropanoic acid
Synonyms: leu-phe
Molecular Weight (g/mol): 278.35
Molecular Formula: C15H22N2O3
InChi Key: KFKWRHQBZQICHA-UHFFFAOYNA-N
SMILES: CC(C)CC(N)C(=O)NC(CC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)