Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Leucyl-DL-phenylalanine 98.0+%, TCI America™
SDP

Catalog No. L00381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L00381G 1 g
L0038100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L00381G Supplier TCI America Supplier No. L00381G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 56217-82-4 | C15H22N2O3 | 278.352 g/mol

Chemical Identifiers

CAS 56217-82-4
Molecular Formula C15H22N2O3
Molecular Weight (g/mol) 278.352
MDL Number MFCD00037258
InChI Key KFKWRHQBZQICHA-UHFFFAOYSA-N
Synonym H-DL-Leu-DL-Phe-OH
PubChem CID 259325
IUPAC Name 2-[(2-amino-4-methylpentanoyl)amino]-3-phenylpropanoic acid
SMILES CC(C)CC(C(=O)NC(CC1=CC=CC=C1)C(=O)O)N

Specifications

Color White
Packaging Glass Bottle
Quantity 1 g
Formula Weight 278.35
Percent Purity ≥98.0% (T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material DL-Leucyl-DL-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.