accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONHSNO

CAS RN 5624-27-1

IUPAC Name: 3-(5-oxo-1-phenyl-2-sulfanylideneimidazolidin-4-yl)propanoic acid
Synonyms: 3-(5-oxo-1-phenyl-2-sulfanylideneimidazolidin-4-yl)propanoic acid
Molecular Weight (g/mol): 264.3
Molecular Formula: C12H12N2O3S
InChi Key: ULKNJYDXLYMGFB-UHFFFAOYNA-N
SMILES: OC(=O)CCC1NC(=S)N(C1=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results