Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylthiohydantoin-glutamic Acid 95.0+%, TCI America™
SDP

Catalog No. P0375100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0375100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0375100MG Supplier TCI America Supplier No. P0375100MG

Specifications

Melting Point 175°C
CAS 5624-27-1
Color White-Yellow
Molecular Formula C12H12N2O3S
MDL Number MFCD00070137
Synonym 3-Phenyl-2-thiohydantoin-5-propionic Acid, PTH-glutamic Acid
InChI Key ULKNJYDXLYMGFB-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)N2C(=O)C(NC2=S)CCC(=O)O
IUPAC Name 3-(5-oxo-1-phenyl-2-sulfanylideneimidazolidin-4-yl)propanoic acid
Quantity 100 mg
Molecular Weight (g/mol) 264.299
PubChem CID 2872155
Formula Weight 264.30
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylthiohydantoin-glutamic Acid
Show More Show Less
RTECSNumber NI9570000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.