accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOSOONNSOHOO

CAS RN 5625-56-9

IUPAC Name: 3-[4-(3-sulfopropyl)piperazin-1-yl]propane-1-sulfonic acid
Synonyms: 3-[4-(3-sulfopropyl)piperazin-1-yl]propane-1-sulfonic acid
Molecular Weight (g/mol): 330.41
Molecular Formula: C10H22N2O6S2
InChi Key: PDLPTSJWDUCMKS-UHFFFAOYSA-N
SMILES: OS(=O)(=O)CCCN1CCN(CCCS(O)(=O)=O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results