accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NCH3CH3H3CH3CI(E/Z)(E/Z)(E/Z)

CAS RN 56289-64-6

IUPAC Name: 1,1,3-trimethyl-2-(5-{1,1,3-trimethyl-1H,2H,3H-benzo[e]indol-2-ylidene}penta-1,3-dien-1-yl)-1H-benzo[e]indol-3-ium iodide
Synonyms: 1,1,3-trimethyl-2-(5-{1,1,3-trimethylbenzo[e]indol-2-ylidene}penta-1,3-dien-1-yl)benzo[e]indol-3-ium iodide
Molecular Weight (g/mol): 610.58
Molecular Formula: C35H35IN2
InChi Key: PXXBSYXTQAWFHV-UHFFFAOYSA-M
SMILES: [I-].CN1C2=CC=C3C=CC=CC3=C2C(C)(C)C1=CC=CC=CC1=[N+](C)C2=CC=C3C=CC=CC3=C2C1(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results