Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IR 676 Iodide 97.0+%, TCI America™
SDP

Catalog No. I10301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10301G 1 g
I1030200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10301G Supplier TCI America Supplier No. I10301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56289-64-6
Molecular Formula C35H35IN2
Molecular Weight (g/mol) 610.583
MDL Number MFCD07433812
InChI Key PXXBSYXTQAWFHV-UHFFFAOYSA-M
Synonym 1,1,3-Trimethyl-2-[5-(1,1,3-trimethyl-1,3-dihydro-2H-benzo[e]indol-2-ylidene)-1,3-pentadienyl]-1H-benzo[e]indolium Iodide
PubChem CID 73181418
IUPAC Name 1,1,3-trimethyl-2-[5-(1,1,3-trimethylbenzo[e]indol-3-ium-2-yl)penta-2,4-dienylidene]benzo[e]indole;iodide
SMILES CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)C)C=CC=CC=C4C(C5=C(N4C)C=CC6=CC=CC=C65)(C)C)C.[I-]

Specifications

Quantity 1 g
Formula Weight 610.58
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material IR 676 Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.