accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 563-12-2

CH3OPSOH3CSSPSOH3COH3C

CAS RN 563-12-2

IUPAC Name: O,O-diethyl [({[diethoxy(sulfanylidene)-λ⁵-phosphanyl]sulfanyl}methyl)sulfanyl]phosphonothioate
Synonyms: ethion diethion caswell no. 427 embathion fosfono 50 rodocid ethopaz ethodan rhodocide ethanox
Molecular Weight (g/mol): 384.46
Molecular Formula: C9H22O4P2S4
InChi Key: RIZMRRKBZQXFOY-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)SCSP(=S)(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results