accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CH3C

CAS RN 563-16-6

IUPAC Name: 3,3-dimethylhexane
Synonyms: 3,3-dimethyl hexane
Molecular Weight (g/mol): 114.23
Molecular Formula: C8H18
InChi Key: KUMXLFIBWFCMOJ-UHFFFAOYSA-N
SMILES: CCCC(C)(C)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results