Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethylhexane 99.0+%, TCI America™
SDP

Catalog No. D12271ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12271ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12271ML Supplier TCI America Supplier No. D12271ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 563-16-6
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00048729
InChI Key KUMXLFIBWFCMOJ-UHFFFAOYSA-N
PubChem CID 11233
IUPAC Name 3,3-dimethylhexane
SMILES CCCC(C)(C)CC

Specifications

Melting Point -126°C
Color Colorless
Boiling Point 112°C
Quantity 1 mL
UN Number 1262
Formula Weight 114.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethylhexane
EINECSNumber (2)-0008
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.