accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 563-58-6

IUPAC Name: 1,1-dichloroprop-1-ene
Synonyms: 1,1-dichloropropene
Molecular Weight (g/mol): 110.97
Molecular Formula: C3H4Cl2
InChi Key: ZAIDIVBQUMFXEC-UHFFFAOYSA-N
SMILES: CC=C(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results