accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOO

CAS RN 5631-96-9

IUPAC Name: 2-(2-chloroethoxy)oxane
Synonyms: 2-(2-chloroethoxy)oxane
Molecular Weight (g/mol): 164.63
Molecular Formula: C7H13ClO2
InChi Key: ZVVQFPGYDBUGQB-UHFFFAOYNA-N
SMILES: ClCCOC1CCCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results