Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Chloroethoxy)tetrahydro-2H-pyran 96.0+%, TCI America™
SDP

Catalog No. C167825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C167825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C167825G Supplier TCI America Supplier No. C167825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5631-96-9
Molecular Formula C7H13ClO2
Molecular Weight (g/mol) 164.629
MDL Number MFCD00142761
InChI Key ZVVQFPGYDBUGQB-UHFFFAOYSA-N
PubChem CID 254951
IUPAC Name 2-(2-chloroethoxy)oxane
SMILES C1CCOC(C1)OCCCl

Specifications

Color Yellow
Boiling Point 104°C
Quantity 25 g
Formula Weight 164.63
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Chloroethoxy)tetrahydro-2H-pyran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.