accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56360-46-4

IUPAC Name: 4',5'-dibromo-2',7'-dinitro-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
Synonyms: 4',5'-dibromo-2',7'-dinitro-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
Molecular Weight (g/mol): 578.08
Molecular Formula: C20H6Br2N2O9
InChi Key: ZBQZBWKNGDEDOA-UHFFFAOYSA-L
SMILES: [O-]C1=C(Br)C2=C(C=C1[N+]([O-])=O)C1(OC(=O)C3=CC=CC=C13)C1=CC(=C([O-])C(Br)=C1O2)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results